accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOBrBr

CAS RN 617-33-4

IUPAC Name: ethyl 2,2-dibromoacetate
Synonyms: ethyl 2,2-dibromoacetate
Molecular Weight (g/mol): 245.9
Molecular Formula: C4H6Br2O2
InChi Key: NIJGVVHCUXNSLL-UHFFFAOYSA-N
SMILES: CCOC(=O)C(Br)Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results