accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 617-27-6

CH3OONH2H3CHCl

CAS RN 617-27-6

IUPAC Name: 1-ethoxy-1-oxopropan-2-aminium chloride
Synonyms: 1-ethoxy-1-oxopropan-2-aminium chloride
Molecular Weight (g/mol): 153.61
Molecular Formula: C5H12ClNO2
InChi Key: JCXLZWMDXJFOOI-UHFFFAOYNA-N
SMILES: [Cl-].CCOC(=O)C(C)[NH3+]
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)