accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 614-99-3

IUPAC Name: ethyl furan-2-carboxylate
Synonyms: ethyl 2-furoate
Molecular Weight (g/mol): 140.14
Molecular Formula: C7H8O3
InChi Key: NHXSTXWKZVAVOQ-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CO1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)