accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3HNH2NS

CAS RN 614-78-8

IUPAC Name: (2-methylphenyl)thiourea
Synonyms: O-tolylthiourea
Molecular Weight (g/mol): 166.24
Molecular Formula: C8H10N2S
InChi Key: ACLZYRNSDLQOIA-UHFFFAOYSA-N
SMILES: CC1=CC=CC=C1NC(N)=S
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results