accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 614-21-1

ONOO

CAS RN 614-21-1

IUPAC Name: 2-nitro-1-phenylethan-1-one
Synonyms: 2-nitroacetophenone
Molecular Weight (g/mol): 165.15
Molecular Formula: C8H7NO3
InChi Key: JTWHVBNYYWFXSI-UHFFFAOYSA-N
SMILES: [O-][N+](=O)CC(=O)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results