accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 61291-91-6

IUPAC Name: (4-methyl-2-phenyl-1,3-thiazol-5-yl)methanol
Synonyms: (4-methyl-2-phenyl-1,3-thiazol-5-yl)methanol
Molecular Weight (g/mol): 205.28
Molecular Formula: C11H11NOS
InChi Key: JSIPFWSPCWZJIU-UHFFFAOYSA-N
SMILES: CC1=C(CO)SC(=N1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results