accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 61150-57-0

FBrBr

CAS RN 61150-57-0

IUPAC Name: 2-bromo-1-(bromomethyl)-4-fluorobenzene
Synonyms: 2-bromo-1-(bromomethyl)-4-fluorobenzene
Molecular Weight (g/mol): 267.92
Molecular Formula: C7H5Br2F
InChi Key: QPLUIZXBWYUFMY-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(CBr)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results