accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 611-32-5

CH3N

CAS RN 611-32-5

IUPAC Name: 8-methylquinoline
Synonyms: 8-methylquinoline ccris 408
Molecular Weight (g/mol): 143.19
Molecular Formula: C10H9N
InChi Key: JRLTTZUODKEYDH-UHFFFAOYSA-N
SMILES: CC1=C2N=CC=CC2=CC=C1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)