accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6108-23-2

LiOO

CAS RN 6108-23-2

IUPAC Name: lithium(1+) formate hydrate
Synonyms: lithium(1+) formate hydrate
Molecular Weight (g/mol): 69.97
Molecular Formula: CH3LiO3
InChi Key: RDAKCPVJSPEDON-UHFFFAOYSA-M
SMILES: [Li+].O.[O-]C=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results