accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6097-07-0

IUPAC Name: 2-(pent-4-yn-1-yl)-2,3-dihydro-1H-isoindole-1,3-dione
Synonyms: 2-(pent-4-yn-1-yl)isoindole-1,3-dione
Molecular Weight (g/mol): 213.24
Molecular Formula: C13H11NO2
InChi Key: YNZIPXLLPFYDGM-UHFFFAOYSA-N
SMILES: O=C1N(CCCC#C)C(=O)C2=CC=CC=C12
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results