accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOONHOOHO

CAS RN 6094-36-6

IUPAC Name: 2-(phenylformamido)pentanedioic acid
Synonyms: 2-(phenylformamido)pentanedioic acid
Molecular Weight (g/mol): 251.24
Molecular Formula: C12H13NO5
InChi Key: LPJXPACOXRZCCP-UHFFFAOYNA-N
SMILES: OC(=O)CCC(NC(=O)C1=CC=CC=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)