accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 60871-84-3

IUPAC Name: nickel(2+) ditrifluoromethanesulfonate
Synonyms: nickel(2+) ditriflate
Molecular Weight (g/mol): 356.82
Molecular Formula: C2F6NiO6S2
InChi Key: KVRSDIJOUNNFMZ-UHFFFAOYSA-L
SMILES: [Ni++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results