accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

OHONONO

CAS RN 6086-24-4

IUPAC Name: 5-carboxy-2,1,3-benzoxadiazol-1-ium-1-olate
Synonyms: 5-carboxy-2,1,3-benzoxadiazol-1-ium-1-olate
Molecular Weight (g/mol): 180.12
Molecular Formula: C7H4N2O4
InChi Key: AXRCJZJVVZUOHN-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC2=NO[N+]([O-])=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results