accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 60836-07-9

H3CCH3H3COHCH3H3C

CAS RN 60836-07-9

IUPAC Name: 2,4,6-trimethylheptan-4-ol
Synonyms: 2,4,6-trimethylheptan-4-ol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: QSVYJSJPLCSACO-UHFFFAOYSA-N
SMILES: CC(C)CC(C)(O)CC(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results