accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 608-05-9

CH3HNOO

CAS RN 608-05-9

IUPAC Name: 5-methyl-2,3-dihydro-1H-indole-2,3-dione
Synonyms: 5-methylisatin
Molecular Weight (g/mol): 161.16
Molecular Formula: C9H7NO2
InChi Key: VAJCSPZKMVQIAP-UHFFFAOYSA-N
SMILES: CC1=CC=C2NC(=O)C(=O)C2=C1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)