accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3CH3OCH3CH3OCo

CAS RN 60406-95-3

IUPAC Name: λ²-cobalt(2+) bis(propan-2-olate)
Synonyms: λ²-cobalt(2+) bis(propan-2-olate)
Molecular Weight (g/mol): 177.11
Molecular Formula: C6H14CoO2
InChi Key: VBXHMSMVQZJJLD-UHFFFAOYSA-N
SMILES: [Co++].CC(C)[O-].CC(C)[O-]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results