accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 6036-48-2

SOOOH

CAS RN 6036-48-2

IUPAC Name: naphthalene-1-sulfonic acid dihydrate
Synonyms: α-naphthalenesulfonic acid dihydrate
Molecular Weight (g/mol): 244.26
Molecular Formula: C10H12O5S
InChi Key: GBWFYBZWCZOBLI-UHFFFAOYSA-N
SMILES: O.O.OS(=O)(=O)C1=CC=CC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results