accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 60230-36-6

IUPAC Name: 2,6-difluorobenzene-1-sulfonyl chloride
Synonyms: 2,6-difluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 212.59
Molecular Formula: C6H3ClF2O2S
InChi Key: QXWAUQMMMIMLTO-UHFFFAOYSA-N
SMILES: FC1=CC=CC(F)=C1S(Cl)(=O)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results