accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 60082-02-2

H3CNO

CAS RN 60082-02-2

IUPAC Name: 1-methyl-2,3-dihydro-1H-indole-5-carbaldehyde
Synonyms: 1-methyl-2,3-dihydroindole-5-carbaldehyde
Molecular Weight (g/mol): 161.2
Molecular Formula: C10H11NO
InChi Key: OZQGVTARHLDSTI-UHFFFAOYSA-N
SMILES: CN1CCC2=C1C=CC(C=O)=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results