accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 60022-62-0

OOOHNH2HCl

CAS RN 60022-62-0

IUPAC Name: benzyl 2-amino-3-hydroxypropanoate hydrochloride
Synonyms: benzyl 2-amino-3-hydroxypropanoate hydrochloride
Molecular Weight (g/mol): 231.68
Molecular Formula: C10H14ClNO3
InChi Key: MGZWCDQAKCHOBX-UHFFFAOYNA-N
SMILES: Cl.NC(CO)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)