accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 60-23-1

NH2HS

CAS RN 60-23-1

IUPAC Name: 2-aminoethane-1-thiol
Synonyms: cysteamine cysteamin β-MEA cysteamide cysteamine bitartate ethanethiolamine cysteamine, 35S-labeled cysteamine, 35S labeled thioethanolamine cysteamine bitartrate preparation
Molecular Weight (g/mol): 77.15
Molecular Formula: C2H7NS
InChi Key: UFULAYFCSOUIOV-UHFFFAOYSA-N
SMILES: NCCS

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results