accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONHN

CAS RN 59843-63-9

IUPAC Name: 5-(2-methoxyphenyl)-1H-pyrazole
Synonyms: 3-(2-methoxyphenyl)-2H-pyrazole
Molecular Weight (g/mol): 174.2
Molecular Formula: C10H10N2O
InChi Key: KLPGJCMICASHKV-UHFFFAOYSA-N
SMILES: COC1=CC=CC=C1C1=CC=NN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results