accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 598-79-8

HOOClH2C

CAS RN 598-79-8

IUPAC Name: 2-chloroprop-2-enoic acid
Synonyms: 2-propenoic acid, 2-chloro-
Molecular Weight (g/mol): 106.51
Molecular Formula: C3H3ClO2
InChi Key: SZTBMYHIYNGYIA-UHFFFAOYSA-N
SMILES: OC(=O)C(Cl)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results