accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3ClO

CAS RN 598-09-4

IUPAC Name: 2-(chloromethyl)-2-methyloxirane
Synonyms: 2-(chloromethyl)-2-methyloxirane
Molecular Weight (g/mol): 106.55
Molecular Formula: C4H7ClO
InChi Key: VVHFXJOCUKBZFS-UHFFFAOYNA-N
SMILES: CC1(CCl)CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results