accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CNNClOOO

CAS RN 59703-00-3

IUPAC Name: 4-ethyl-2,3-dioxopiperazine-1-carbonyl chloride
Synonyms: 4-ethyl-2,3-dioxopiperazine-1-carbonyl chloride
Molecular Weight (g/mol): 204.61
Molecular Formula: C7H9ClN2O3
InChi Key: SXVBQOZRZIUHKU-UHFFFAOYSA-N
SMILES: CCN1CCN(C(Cl)=O)C(=O)C1=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results