accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 59588-86-2

CH3H3C(S)HNOHNOH3CCH3HNO(S)NHOCH3H3CNHO(S)NH2OHNO(S)H3CHNOH3CCH3HNO(S)H3CNHOH3CCH3NHO(S)NOCH3H3CNHCH3OH3CCH3OHNH3CCH3ON(S)OHN(S)H3CCH3OHNH3CCH3OHNH3CCH3OHN(S)HOOOHN(S)H2NOONH(S)HO

CAS RN 59588-86-2

IUPAC Name: (4S)-4-{[(1S)-3-carbamoyl-1-{[(2S)-1-hydroxy-3-phenylpropan-2-yl]carbamoyl}propyl]carbamoyl}-4-(2-{2-[(2S)-2-{[(2S)-1-{2-[(2S)-2-(2-{2-[(2S)-2-{2-[(2S)-4-carbamoyl-2-[(2S)-2-{2-[(2S)-2-(2-{[(2S)-1-(2-acetamido-2-methylpropanoyl)pyrrolidin-2-yl]formamido}-2-methylpropanamido)propanamido]-2-methylpropanamido}propanamido]butanamido]-2-methylpropanamido}-3-methylbutanamido]-2-methylpropanamido}acetamido)-4-methylpentanamido]-2-methylpropanoyl}pyrrolidin-2-yl]formamido}-3-methylbutanamido]-2-methylpropanamido}-2-methylpropanamido)butanoic acid
Synonyms: alamethicin
Molecular Weight (g/mol): 1964.34
Molecular Formula: C92H150N22O25
InChi Key: LGHSQOCGTJHDIL-UTXLBGCNSA-N
SMILES: CC(C)C[C@H](NC(=O)CNC(=O)C(C)(C)NC(=O)[C@@H](NC(=O)C(C)(C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](C)NC(=O)C(C)(C)NC(=O)[C@H](C)NC(=O)C(C)(C)NC(=O)[C@@H]1CCCN1C(=O)C(C)(C)NC(C)=O)C(C)C)C(=O)NC(C)(C)C(=O)N1CCC[C@H]1C(=O)N[C@@H](C(C)C)C(=O)NC(C)(C)C(=O)NC(C)(C)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@H](CO)CC1=CC=CC=C1
Color 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Type 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (3)