accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 59516-96-0

CH3OOOBr

CAS RN 59516-96-0

IUPAC Name: methyl 3-(2-bromoethoxy)benzoate
Synonyms: methyl 3-(2-bromoethoxy)benzoate
Molecular Weight (g/mol): 259.1
Molecular Formula: C10H11BrO3
InChi Key: CGGLKWTURIUCMN-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(OCCBr)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results