accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3OON(S)OO

CAS RN 59378-75-5

IUPAC Name: (3S)-1-[(tert-butoxy)carbonyl]pyrrolidine-3-carboxylate
Synonyms: (3S)-1-(tert-butoxycarbonyl)pyrrolidine-3-carboxylate
Molecular Weight (g/mol): 214.24
Molecular Formula: C10H16NO4
InChi Key: HRMRQBJUFWFQLX-ZETCQYMHSA-M
SMILES: CC(C)(C)OC(=O)N1CC[C@@H](C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results