accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 593-96-4

CH3ClBr

CAS RN 593-96-4

IUPAC Name: 1-bromo-1-chloroethane
Synonyms: 1-bromo-1-chloroethane
Molecular Weight (g/mol): 143.41
Molecular Formula: C2H4BrCl
InChi Key: QMSVNDSDEZTYAS-UHFFFAOYNA-N
SMILES: CC(Cl)Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)