CAS RN 593-08-8
CAS RN 593-08-8
IUPAC Name:
tridecan-2-one
Synonyms:
2-tridecanone
Molecular Weight (g/mol):
198.35
Molecular Formula:
C13H26O
InChi Key:
CYIFVRUOHKNECG-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCCC(C)=O
1
–
1
of
1
results
1
2-Tridecanone, 98+%
CAS: 593-08-8 Molecular Formula: C13H26O Molecular Weight (g/mol): 198.35 MDL Number: MFCD00008968 InChI Key: CYIFVRUOHKNECG-UHFFFAOYSA-N Synonym: 2-tridecanone,methyl undecyl ketone,mathyl undecyl kepoje,hendecyl methyl ketone,2-tridecankje,tridecanone-2,methyl n-undecyl ketone,tridecanone,2-tridecanone natural,unii-5q35vhx26k PubChem CID: 11622 ChEBI: CHEBI:77928 IUPAC Name: tridecan-2-one SMILES: CCCCCCCCCCCC(=O)C