accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 59214-70-9

OBr

CAS RN 59214-70-9

IUPAC Name: 3-bromo-1-benzofuran
Synonyms: 3-bromo-1-benzofuran
Molecular Weight (g/mol): 197.03
Molecular Formula: C8H5BrO
InChi Key: ICJNAOJPUTYWNV-UHFFFAOYSA-N
SMILES: BrC1=COC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results