accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COONNCl

CAS RN 59128-02-8

IUPAC Name: ethyl 2-{6-chloroimidazo[1,2-a]pyridin-2-yl}acetate
Synonyms: ethyl 2-{6-chloroimidazo[1,2-a]pyridin-2-yl}acetate
Molecular Weight (g/mol): 238.67
Molecular Formula: C11H11ClN2O2
InChi Key: JZTURCHIBDOZFO-UHFFFAOYSA-N
SMILES: CCOC(=O)CC1=CN2C=C(Cl)C=CC2=N1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results