accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 59122-55-3

H3CO(R)O(R)OH(S)OH(S)OH(R)HO

CAS RN 59122-55-3

IUPAC Name: (2R,3R,4S,5S,6R)-2-(dodecyloxy)-6-(hydroxymethyl)oxane-3,4,5-triol
Synonyms: lauryl glucoside
Molecular Weight (g/mol): 348.48
Molecular Formula: C18H36O6
InChi Key: PYIDGJJWBIBVIA-UYTYNIKBSA-N
SMILES: CCCCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results