CAS RN 5910-89-4
IUPAC Name:
2,3-dimethylpyrazine
Synonyms:
pyrazine, 2,3-dimethyl-
Molecular Weight (g/mol):
108.14
Molecular Formula:
C6H8N2
InChi Key:
OXQOBQJCDNLAPO-UHFFFAOYSA-N
SMILES:
CC1=NC=CN=C1C
1
–
3
of
3
results
1
2,3-Dimethylpyrazine, 99%
CAS: 5910-89-4 Molecular Formula: C6H8N2 Molecular Weight (g/mol): 108.14 MDL Number: MFCD00006144 InChI Key: OXQOBQJCDNLAPO-UHFFFAOYSA-N Synonym: pyrazine, 2,3-dimethyl,2,3-dimethyl-1,4-diazine,2,3-dimethyl-pyrazine,pyrazine, dimethyl,2,3-dimethyl pyrazine,ccris 2928,unii-whf7883d0v,fema no. 3271,natural,dimethylpyrazine PubChem CID: 22201 IUPAC Name: 2,3-dimethylpyrazine SMILES: CC1=NC=CN=C1C
3
2,3-Dimethylpyrazine, TRC
CAS: 5910-89-4 Molecular Formula: C6H8N2 Molecular Weight (g/mol): 108.14 Synonym: 2,3-Dimethylpyrazine,Pyrazine, 2,3-dimethyl- (6CI, 7CI, 8CI, 9CI, ACI) SMILES: Cc1nccnc1C