accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5908-81-6

BaOHOHOOOO

CAS RN 5908-81-6

IUPAC Name: barium(2+) 2,3-dihydroxybutanedioate
Synonyms: barium(2+) tartrate
Molecular Weight (g/mol): 285.4
Molecular Formula: C4H4BaO6
InChi Key: HQYOWPDUMCBSLQ-UHFFFAOYNA-L
SMILES: [Ba++].OC(C(O)C([O-])=O)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results