accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

FNCOF

CAS RN 59025-55-7

IUPAC Name: 2,4-difluoro-1-isocyanatobenzene
Synonyms: 2,4-difluoro-1-isocyanatobenzene
Molecular Weight (g/mol): 155.1
Molecular Formula: C7H3F2NO
InChi Key: HNENEALJPWJWJY-UHFFFAOYSA-N
SMILES: FC1=CC=C(N=C=O)C(F)=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results