accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 590-50-1

CH3OCH3CH3H3C

CAS RN 590-50-1

IUPAC Name: 4,4-dimethylpentan-2-one
Synonyms: 2-pentanone, 4,4-dimethyl-
Molecular Weight (g/mol): 114.19
Molecular Formula: C7H14O
InChi Key: AZASWMGVGQEVCS-UHFFFAOYSA-N
SMILES: CC(=O)CC(C)(C)C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)