accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H2OH2OCuOHOHOOCuOHOHOO

CAS RN 5893-66-3

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 343.19
Molecular Formula: C4H8Cu2O10
InChi Key: NNZNHNMQOYUANB-UHFFFAOYSA-N
SMILES: O.O.[Cu].[Cu].OC(=O)C(O)=O.OC(=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results