accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H(S)(R)(S)NO(S)NOCH3OH3COH(R)(R)HHHH2O4

CAS RN 5892-11-5

IUPAC Name: (1R,11S,18S,20R,21R,22S)-4,5-dimethoxy-12-oxa-8,17-diazaheptacyclo[15.5.2.0¹,¹⁸.0²,⁷.0⁸,²².0¹¹,²¹.0¹⁵,²⁰]tetracosa-2(7),3,5,14-tetraen-9-one tetrahydrate
Synonyms: brucine tetrahydrate
Molecular Weight (g/mol): 466.53
Molecular Formula: C23H34N2O8
InChi Key: XLXHHXCMBNBDMP-BEYGIVOKSA-N
SMILES: O.O.O.O.[H][C@]12C[C@@]3([H])C4=CCO[C@@]5([H])CC(=O)N6C7=C(C=C(OC)C(OC)=C7)[C@@]1(CCN2C4)[C@]6([H])[C@@]35[H]
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results