accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COONNH3

CAS RN 58859-46-4

IUPAC Name: 1-(ethoxycarbonyl)piperidin-4-aminium
Synonyms: 1-(ethoxycarbonyl)piperidin-4-aminium
Molecular Weight (g/mol): 173.24
Molecular Formula: C8H17N2O2
InChi Key: GQQQULCEHJQUJT-UHFFFAOYSA-O
SMILES: CCOC(=O)N1CCC([NH3+])CC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)