accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 58775-07-8

CH3CH3H3CH3CH3CCH3

CAS RN 58775-07-8

IUPAC Name: 3,6-di-tert-butyl-9H-fluorene
Synonyms: 3,6-di-tert-butyl-9H-fluorene
Molecular Weight (g/mol): 278.44
Molecular Formula: C21H26
InChi Key: IWUHFBNEZZKUEY-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC2=C(CC3=C2C=C(C=C3)C(C)(C)C)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results