accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 58749-51-2

SNCO

CAS RN 58749-51-2

IUPAC Name: 2-(2-isocyanatoethyl)thiophene
Synonyms: 2-(2-isocyanatoethyl)thiophene
Molecular Weight (g/mol): 153.2
Molecular Formula: C7H7NOS
InChi Key: KAJZMNLREUEZSH-UHFFFAOYSA-N
SMILES: O=C=NCCC1=CC=CS1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results