accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 5866-98-8

IUPAC Name: 2,6-dichloro-3-nitrobenzonitrile
Synonyms: 2,6-dichloro-3-nitrobenzonitrile
Molecular Weight (g/mol): 217.01
Molecular Formula: C7H2Cl2N2O2
InChi Key: NSKVWZIEYFSHIM-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C(Cl)C(C#N)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results