accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 58626-38-3

IUPAC Name: 2,5-dioxopyrrolidin-1-yl 3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)benzoate
Synonyms: 2,5-dioxopyrrolidin-1-yl 3-(2,5-dioxopyrrol-1-yl)benzoate
Molecular Weight (g/mol): 314.25
Molecular Formula: C15H10N2O6
InChi Key: LLXVXPPXELIDGQ-UHFFFAOYSA-N
SMILES: O=C(ON1C(=O)CCC1=O)C1=CC=CC(=C1)N1C(=O)C=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results