CAS RN 5851-43-4
IUPAC Name:
2-(propan-2-yl)-1H-1,3-benzodiazole
Synonyms:
2-isopropyl-1H-1,3-benzodiazole
Molecular Weight (g/mol):
160.22
Molecular Formula:
C10H12N2
InChi Key:
RITUGMAIQCZEOG-UHFFFAOYSA-N
SMILES:
CC(C)C1=NC2=CC=CC=C2N1
1
–
1
of
1
results
1
Alfa Aesar™ 2-Isopropylbenzimidazole, 98%
CAS: 5851-43-4 Molecular Formula: C10H12N2 Molecular Weight (g/mol): 160.22 MDL Number: MFCD00051759 InChI Key: RITUGMAIQCZEOG-UHFFFAOYSA-N Synonym: 2-isopropyl-1h-benzo d imidazole,2-isopropylbenzimidazole,2-isopropyl-1h-benzimidazole,benzimidazole, 2-isopropyl,1h-benzimidazole, 2-1-methylethyl,2-propan-2-yl-1h-1,3-benzodiazole,2-isopropyl-1h-1,3-benzodiazole,1h-benzimidazole, 2-1-methylethyl-9ci,2-methylethyl benzimidazole,acmc-1avkb PubChem CID: 22121 IUPAC Name: 2-propan-2-yl-1H-benzimidazole SMILES: CC(C)C1=NC2=CC=CC=C2N1