accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CNH2

CAS RN 585-32-0

IUPAC Name: 2-phenylpropan-2-amine
Synonyms: dimethyl-aminophenylmethane
Molecular Weight (g/mol): 135.21
Molecular Formula: C9H13N
InChi Key: KDFDOINBXBEOLZ-UHFFFAOYSA-N
SMILES: CC(C)(N)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results