accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

HOONI

CAS RN 58481-10-0

IUPAC Name: 2-iodopyridine-4-carboxylic acid
Synonyms: 2-iodopyridine-4-carboxylic acid
Molecular Weight (g/mol): 249.01
Molecular Formula: C6H4INO2
InChi Key: FALCXGCLOUKBSP-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=NC(I)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results