accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 58419-69-5

BrNNN

CAS RN 58419-69-5

IUPAC Name: 1-[4-(bromomethyl)phenyl]-1H-1,2,4-triazole
Synonyms: 1-[4-(bromomethyl)phenyl]-1,2,4-triazole
Molecular Weight (g/mol): 238.09
Molecular Formula: C9H8BrN3
InChi Key: NOUAEJFAVOHNSY-UHFFFAOYSA-N
SMILES: BrCC1=CC=C(C=C1)N1C=NC=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results