accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ONONHNFFF

CAS RN 58315-38-1

IUPAC Name: 1-[2-nitro-4-(trifluoromethyl)phenyl]piperazine
Synonyms: 1-[2-nitro-4-(trifluoromethyl)phenyl]piperazine
Molecular Weight (g/mol): 275.23
Molecular Formula: C11H12F3N3O2
InChi Key: YOBUPGXTLFRIJD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(=CC=C1N1CCNCC1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results