accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 583-71-1

CH3BrH3C

CAS RN 583-71-1

IUPAC Name: 4-bromo-1,2-dimethylbenzene
Synonyms: benzene, 4-bromo-1,2-dimethyl-
Molecular Weight (g/mol): 185.06
Molecular Formula: C8H9Br
InChi Key: QOGHRLGTXVMRLM-UHFFFAOYSA-N
SMILES: CC1=CC=C(Br)C=C1C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)